Learn More
Ebro Electronic™ Datalogger serie EBI 20
Descrizione
- Connected to the PC and supplied with power from there by means of a USB cable
- Supplied with Interface, USB cable, software Winlog.basic, instruction manual
- Clean with a slightly damp cloth only
Order Info
Interface
Specifica
Specifica
| Tipo | Mounting Holder |
| Tipo di display | No Display |
| Calibrazione | Not Calibrated |
| Tipi di allarme | None |
| Interfaccia | EBI 20-WM |
| Modello | EBI 20-WM |