Learn More
Thermo Scientific™ Accessories and Kits for Orion™ 2111LL Low Level Sodium Analyzers
Codice prodotto. 16864531
Cambia vista
Click to view available options
Tipo:
Tubing Kit
Dimensione della confezione:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Dimensione della confezione
16864531 Tubing Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16864531 Fornitore Thermo Scientific™ N. del fornitore 211194
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
missing translation for 'validMainframeMsg'
Have all the auxiliary items on hand for top production with these accessories and kits.
Have all the auxiliary items on hand for top production. We offer an assortment of accessories and kits for the Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzers. Accessories and kits include calibration kit and carrrying case, analyzer enclosure, service kit, maintenance kit, consumable kit, and related electrodes.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Diffusion Tubing Kit - includes 4 inch diffusion tubing assembly with O-rings, for use with DIPA reagent used on 2111LL, 2111XP, 1811EL analyzers
Tipo Tubing Kit
Da utilizzare con (apparecchiatura) Orion™ 2111LL Low Level Sodium Analyzers
Include 4 inch diffusion tubing assembly with O-rings, for use with DIPA reagent used on 2111LL, 2111XP, 1811EL analyzers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.