Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in a wide range of applications and in conjunction with our various lines of filtration devices.
Marca:  Thermo Scientific™ 710300-0001
23.90 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.01200kg
Codice prodotto. 10182541
  • 35.85€ / Pezzo
Spedizione stimata: 12-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Features
  • Graduated receiver allows easy visualization of sample volume
  • Two side arms allow connection to vacuum line
  • Upper chamber is designed for pressure or vacuum filtration
  • Independent locking rings seal upper chamber to receiver without damagingmembrane to ensure efficient filtration
  • Chamber accommodates 47 or 50mmmembranes (not included) and has three ports for venting (with or without syringe filters) and aseptic addition of samples
  • Ports also can be attached to a pressure source
  • Upper chamber is supplied with twomembrane support plates: a sterilization plate and an analytical plate
Receiver Accepts
  • 0.25 to 0.3125 in. I.D. (6.4 to 8mm) vacuum tubing
  • Syringe filters
Sterilization Plate (one-piece, clear)
  • Providesmaximum flowrate and throughput
  • Suitable for cold sterilization or clarification of culturemedia, reagents or solvents that are compatible with holder andmembranematerials
  • Nominal filter area, 13.3cm2
Analytical Plate (two-piece, white)
  • Provides optimalmembrane support and keepsmembrane flat to facilitate analytical procedures
  • Suitable for analysis of particulates and biological contaminants, or anymicrobiological analyses using themembrane filter technique
  • Nominal filter area, 11.3cm2
Includes:
Complete upper chamber, receiver, polypropylene cover and TPE port caps. (membranes not included.)
Specifica
Specifica
O-Rings and Gaskets