Learn More
Merck Millipore Chemicon™ Accutase™ Cell Detachment Solution
useful for creating single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue.
Marca: Merck Millipore SCR005
Dettagli aggiuntivi : Peso : 0.01350kg