Learn More
Acebutolol EP Impurity F, Mikromol™
Codice prodotto. 30050522
Cambia vista
Click to view available options
Quantità:
25 mg
Dimensione della confezione:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30050522 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30050522 Fornitore Mikromol N. del fornitore MM0435.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Acebutolol EP Impurity F
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico N-[3-Acetyl-4-[(2RS)-2,3-dihydroxypropoxy]phenyl]butanamide
CAS 96480-91-0
Nomi componenti o analita N-[3-Acetyl-4-[(2RS)-2,3-dihydroxypropoxy]phenyl]butanamide
Shipping Condition Room Temperature
Formula molecolare C15 H21 N O5
Certificazione/Conformità ISO 9001
Sinonimo Acebutolol Hydrochloride Imp. F (EP), Acebutolol Imp. F (EP), N-[3-Acetyl-4-[(2RS)-2,3-dihydroxypropoxy]phenyl]butanamide, Acebutolol Hydrochloride Impurity F, Acebutolol Impurity F
InChI Formula InChI=1S/C15H21NO5/c1-3-4-15(20)16-11-5-6-14(13(7-11)10(2)18)21-9-12(19)8-17/h5-7,12,17,19H,3-4,8-9H2,1-2H3,(H,16,20)
SMILES CCCC(=O)Nc1ccc(OCC(O)CO)c(c1)C(=O)C
IUPAC Name N-[3-acetyl-4-(2,3-dihydroxypropoxy)phenyl]butanamide
Molecular Weight (g/mol) 295.33
Quantità 25 mg
Peso formulazione 295.142 g/mol