missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Click to view available options
Quantità:
5 mg
25 mg
Descrizione
A carboxyrhodamine 110 based labeling reagent; reacts with azides via a copper-catalyzed click reactions. Carboxyrhodamine 110 is a bright and photostable green fluorescent label that can be excited at a wavelength of 488nm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Materiale o nome chimico | Acetylene-PEG4-carboxyrhodamine 110 conjugate |
Colore | Red |
Quantità | 5 mg |
Informazioni di solubilità | Soluble in DMSO,DMF,or methanol |
Peso formulazione | 587.63 |
Forma fisica | Solid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'