Learn More
Acriflavine Hydrochloride (>90%), TRC
Product Code. 30132536
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132536 10 g 10g
30138734 1 g 1g
30007740 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132536 Supplier Toronto Research Chemicals Supplier No. TRCA19109010G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 69235-50-3
Molecular Formula C27H27Cl3N6
Molecular Weight (g/mol) 541.9
Synonym Acriflavine Dihydrochloride, Acriflavinium Chloride, Flavacridine Hydrochloride, NSC 689003
IUPAC Name acridine-3,6-diamine;10-methylacridin-10-ium-3,6-diamine;chloride;dihydrochloride
SMILES NC1=CC2=C(C=C1)C=C3C(C=C(N)C=C3)=[N+]2C.NC4=CC5=C(C=C4)C=C6C(C=C(N)C=C6)=N5.[Cl-]
Tekniske data
Quantity 10 g
InChI Formula InChI=1S/C14H13N3.C13H11N3.3ClH/c1-17-13-7-11(15)4-2-9(13)6-10-3-5-12(16)8-14(10)17;14-10-3-1-8-5-9-2-4-11(15)7-13(9)16-12(8)6-10;;;/h2-8H,1H3,(H3,15,16);1-7H,14-15H2;3*1H
Formula Weight 540.1363
Chemical Name or Material Acriflavine Hydrochloride
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.