Learn More
Cytiva Acrodisc™ PF Syringe Filters with Supor™ Membrane, Sterile
Descrizione
- Syringe filters perform sample clean-up and preparation, ensuring the purest samples are analyzed in liquid chromatography, mass spectrometry, and ion chromatography applications
- Superior flow rates and higher throughputs than competitive devices
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
| Materiale (membrana) | PES |
| Dimensione del poro | 0.8/0.2 μm |
| Sterilità | Sterile |
| Quantità | 50/Pk. |
| Diametro (sistema metrico) | 32 mm |