Learn More
Cytiva AcroPak™ 400 Capsules with Supor™ EKV Membrane
Codice prodotto. 17194231
Cambia vista
Click to view available options
Stile uscita:
1 to 1.5 in. Sanitary Flange
1/2 in. Hose Barb Inlet with Filling Bell on Outlet
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Stile uscita unitSize
17194231 1/2 in. Hose Barb Inlet with Filling Bell on Outlet Pezzo
17184231 1 to 1.5 in. Sanitary Flange Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 17194231 Fornitore Cytiva N. del fornitore 12461
  • ONLINE EXCLUSIVE PRICE
    149.00€
    137.81€ / pezzi 1
    Risparmia 11.19€
    7% Sconto
Visualizza tutte le promozioni
In stock
Aggiungi al carrello
Aggiungi al carrello
AcroPak capsules designed for research applications only.
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifica
Materiale (membrana) Hydrophilic PES
Dimensione del poro 0.2 μm
Sterilità Sterile
Area di filtraggio 375 cm2
Stile uscita 1/2 in. Hose Barb Inlet with Filling Bell on Outlet
Quantità 1/Ea.