Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
| Materiale (membrana) | Supor (PES) |
| Dimensione del poro | 0.2 μm |
| Sterilità | Non-sterile |
| Materiale alloggiamento | Polypropylene Housing |
| Quantità | 10/Pk. |
| Volume (sistema metrico) | 350 μL |