Learn More
- Home
- Products
- Baths
- Bath Accessories
- HUBER Adattatore per bagno
HUBER Adattatore per bagno
Codice prodotto. 10759562
Cambia vista
Click to view available options
Size:
M16 x 1 female to G 1/2 female
M16 x 1 female to G 3/4 female
M16 x 1 female to M16 x 1 female
M16 x 1 female to M30 x 1.5 male
M16 x 1 female to R 1/2 male
M16 x 1 male to G1/2 female
M16 x 1 male to G1/2 male
M16 x 1 male to G3/4 female
M16 x 1 male to M16 x 1 male
M16 x 1 male to M24 x 1,5 female
M16 x 1 male to M24 x 1.5 male
M16 x 1 male to M30 x 1.5 female
M24 x 1.5 female to 3/4 NPT female
M24 x 1.5 male M30 x 1.5 female
M24 x 1.5 male to RP 1/2 female
M30 x 1.5 female to G 1 female
M30 x 1.5 female to G 3/8 male
M30 x 1.5 female to G1/2 female
M30 x 1.5 female to G1/2 male
M30 x 1.5 female to G3/4 female
M30 x 1.5 female to M38 x 1.5 male
M30 x 1.5 male to G 1 male
M30 x 1.5 male to G 1/2 male
M30 x 1.5 male to G3/4 female
M30 x 1.5 male to Rp1/2 female
Rp1/2 female to 3/4 NPT female
Rp1/2 female to Rp1/2 female
Dimensione della confezione:
Pezzo
27 product options available for selection
| Codice prodotto. | Size | unitSize |
|---|---|---|
| 10759562 | Rp1/2 female to Rp1/2 female | Pezzo |
| 10638772 | Rp1/2 female to 3/4 NPT female | Pezzo |
| 10127651 | M16 x 1 female to R 1/2 male | Pezzo |
| 10640303 | M16 x 1 female to G 3/4 female | Pezzo |
| 10209511 | M16 x 1 female to G 1/2 female | Pezzo |
| 10259651 | M16 x 1 female to M16 x 1 female | Pezzo |
| 10584922 | M16 x 1 female to M30 x 1.5 male | Pezzo |
| 10504552 | M16 x 1 male to G1/2 female | Pezzo |
| 10127701 | M16 x 1 male to G1/2 male | Pezzo |
| 10289551 | M16 x 1 male to G3/4 female | Pezzo |
| 10341732 | M16 x 1 male to M16 x 1 male | Pezzo |
| 10750984 | M24 x 1.5 female to 3/4 NPT female | Pezzo |
| 10243722 | M16 x 1 male to M30 x 1.5 female | Pezzo |
| 10023692 | M16 x 1 male to M24 x 1,5 female | Pezzo |
| 11557384 | M24 x 1.5 male M30 x 1.5 female | Pezzo |
| 10013742 | M16 x 1 male to M24 x 1.5 male | Pezzo |
| 10783276 | M24 x 1.5 male to RP 1/2 female | Pezzo |
| 10025661 | M30 x 1.5 female to G 1 female | Pezzo |
| 10219701 | M30 x 1.5 female to G1/2 female | Pezzo |
| 10600693 | M30 x 1.5 female to G1/2 male | Pezzo |
| 10127941 | M30 x 1.5 female to G3/4 female | Pezzo |
| 10670493 | M30 x 1.5 female to G 3/8 male | Pezzo |
| 10299741 | M30 x 1.5 male to G 1 male | Pezzo |
| 10209701 | M30 x 1.5 male to Rp1/2 female | Pezzo |
| 10197741 | M30 x 1.5 male to G 1/2 male | Pezzo |
| 10289741 | M30 x 1.5 male to G3/4 female | Pezzo |
| 10509625 | M30 x 1.5 female to M38 x 1.5 male | Pezzo |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Codice prodotto.
10759562
Fornitore
HUBER
N. del fornitore
6358
Accedi o Registrati per verificare il tuo prezzo e la disponibilità.
-
39.50€
37.09€ / pezzi 1
Risparmia 2.41€
6% Sconto
Spedizione stimata: 15-09-2026
Eseguire il login per visualizzare lo stock disponibile
Eseguire il login per visualizzare lo stock disponibile
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit.