Learn More
Motic™ Adapter, Brightfield, LM Plan
Codice prodotto. 15445332
Cambia vista
Click to view available options
Da utilizzare con (apparecchiatura):
LM Plan Objective, AE2000MET Microscopes
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con (apparecchiatura) unitSize
15445332 LM Plan Objective, AE2000MET Microscopes Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15445332 Fornitore Motic™ N. del fornitore 1101010400011
 Articolo soggetto a restrizioni
Spedizione stimata: 25-09-2026
Aggiungi al carrello
Aggiungi al carrello
Accommodate a brightfield LED with this adapter. Motic™ Adapter, Brightfield, LM Plan is specifically designed for LM Plan objectives.
  • Accommodates brightfield LED.
  • For LM Plan objective
  • Leverage advantages of brightfield microscopy:
    • Optics do not change observed structure color.
    • Optics do not change stain colors.
    • Generally less expensive than phase contrast optics.
    • Requires fewer adjustments for specimen observation.
Specifica
Tipo Bright Field
Da utilizzare con (apparecchiatura) LM Plan Objective, AE2000MET Microscopes