Learn More
Fisherbrand™ Adapter for Traceable™ Meters, Universal VAC to 9 VDC
Product Code. 17337629 Shop All Cole-Parmer Products
Change view
Click to view available options
For Use With (Equipment):
Traceable™ Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17337629 Traceable™ Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17337629 Supplier Fisherbrand™ Supplier No. 1960134
  • ONLINE EXCLUSIVE PRICE
    61.95€
    47.84€ / pezzi 1
    Risparmia 14.11€
    22% Sconto
Spedizione stimata: 22-01-2027
Optimize the performance of your conductivity and pH/ORP meters with the Adapter for Traceable™ Meters, Universal VAC to 9 VDC.
  • Includes four different power receptacle adapters to convert 115/220 VAC to 9 VDC
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Type Power Adapter
Certifications/Compliance None
For Use With (Equipment) Traceable™ Meters