Learn More
Eppendorf™ Adapters For Round Bucket in Rotor A-4-38
Product Code. 11883882 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1.5 to 2 mL
15 mL
2.6 to 7 mL
25 mL
85 mL
9 to 15 mL
Tube Type:
Conical Falcon
Eppendorf
Monovette
Oak Ridge
Round Bottom
Standard
Vacutainer
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Tube Type Unit Size
11883882 15 mL Round Bottom Pack of 2
11512215 2.6 to 7 mL Vacutainer Pack of 2
10402652 1.5 to 2 mL Eppendorf Pack of 2
10659651 85 mL Oak Ridge Pack of 2
10574832 25 mL Standard Pack of 2
11572215 9 to 15 mL Monovette Pack of 2
10166461 15 mL Conical Falcon Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11883882 Supplier Eppendorf™ Supplier No. 5702735000
  • ONLINE EXCLUSIVE PRICE
    212.00€
    193.00€ / pezzi 2
    Risparmia 19.00€
    8% Sconto
Spedizione stimata: 09-12-2026
Wide variety of adapters for all blood collection tubes and cell culture tube types
  • Adapter for Rotor A-4-38
  • For centrifugation of blood collection tubes or conical tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type Round Bottom
Capacity 15 mL
For Use With (Equipment) Round bucket in Rotor A-4-38
Bore Size (Metric) 16 mm
No. of Places 16
No. per Pack 2 Pack