Learn More
Adenosine 5'-Diphosphate Sodium Salt, TRC
Product Code. 30325007
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325007 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30325007 Supplier Toronto Research Chemicals Supplier No. TRCA28170825G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Adenosine 5'-Diphosphate Sodium Salt
CAS 20398-34-9
Purity Grade Notes HPLC
Molecular Formula C10H15N5O10P2 . x(Na)
Quantity 25 g
Synonym Adenosine 5-Diphosphate Monosodium Salt
InChI Formula InChI=1S/C10H15N5O10P2/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1
SMILES [Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)[O-])[C@@H](O)[C@H]3O
IUPAC Name [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate
Molecular Weight (g/mol) 427.2
Formula Weight 449.0114
Percent Purity >95