missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Triphosphate Disodium Salt Hydrate MP Biomedicals

Catalog Number 11403091
Click to view available options
Quantità:
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
51963-61-2
C10H20N5Na2O16P3
605.19
MFCD00150754
MWEQTWJABOLLOS-MVHBFMHHNA-L
134129625
O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
51963-61-2
C10H20N5Na2O16P3
605.19
MFCD00150754
MWEQTWJABOLLOS-MVHBFMHHNA-L
134129625
O.O.O.[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Adenosine-5'-triphosphate Disodium Salt Hydrate Cell culture reagent, ≥96%, White Powder

  • Adenosine 5'-triphosphate (ATP) and its phosphate bonds are the basic components of energy exchange in many biological systems.
Materiale o nome chimico Adenosine-5'-Triphosphate Disodium Salt Hydrate
Nota nome Hydrate
Colore White
Forma fisica Powder
Quantità 5 g
Sinonimo adenosine 5'-triphosphate disodium salt trihydrate, adenosine 5'-triphosphate disodium salt, adenosine 5'-triphosphate disodium salt trihydrate, disodium salt
IUPAC Name disodium;[[[(2R,3S,4S,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl]oxy-oxidophosphoryl] hydrogen phosphate;trihydrate
Peso formulazione 551.2 (Anhydrous)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'