missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AdipoRon hydrochloride

Codice prodotto. 18737118
Cambia vista
Click to view available options
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
18737118 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18737118 Fornitore R&D Systems™ N. del fornitore 5096/10
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

A Tocris product, part of R&D Systems. Orally active AdipoR1 and AdipoR2 agonist

  • Agonist of adiponectin receptors AdipoR1 and AdipoR2 (Kd values are 1.8 and 3.1 μm respectively).
  • Activates AMPK and PPARα signaling; ameliorates insulin resistance, dyslipidemia and glucose intolerance in db/db mice.
  • Orally active antidiabetic agent.
TRUSTED_SUSTAINABILITY

Specifica

Materiale o nome chimico 2-(4-Benzyoylphenoxy)-N-[1-(phenylmethyl)-4-piperidinyl]acetamide hydrochloride
CAS 1781835-20-8
Quantità 10 mg
Linea di prodotti Tocris
Formula molecolare C27H28N2O3.HCl
Purezza 98%
Titolo del prodotto
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.