Learn More
Vilbar Lourmat BIO1D SOFTWARE
Product Code. 11761159
Change view
Click to view available options
Product Type:
Administrator Software CFR21
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11761159 Administrator Software CFR21 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11761159 Supplier Vilbar Lourmat Supplier No. 141010011
  • ONLINE EXCLUSIVE PRICE
    1615.00€
    1384.39€ / pezzi 1
    Risparmia 230.61€
    14% Sconto
Spedizione stimata: 08-01-2027
Administrator software ideal for calculating automatic band detection and molecular weight.
  • Pentium, 3.2 GHz, FSB 800 MHz, 33 MHz PCI bus, 512 MB Ram, 10 GB hard disk with at least 100 MB of free space for software installation and image backup, 1280 x 1024 VGA card in 16 million colors (24 Bits), and at least 1 PCI port and 1 free USB port
  • Bio1-D software
  • Transformation of the 1D image into a 3D graph for quantification of the bands
  • Automatic band detection and molecular weight calculation
  • Simultaneous display of the gel and the values ​​of each strip on the same screen
  • Recalculation of values ​​according to a reference calibration curve
  • Quantitative analysis of microplates
  • Colony count according to their characteristics (volume, area, perimeter)
  • Ieporting integrating images of gels, result tables, curves that can be printed or exported in a Windows ™ compatible format
  • Full GLP compliance
Specifications
Product Type Administrator Software CFR21