Learn More
Grant Instruments™ Advanced Air Cleaner Recirculator, 2 Lamps
Descrizione
The inner mirror surface of the recirculator chamber reflects ultraviolet rays thereby increasing the density of UV radiation and enhancing the disinfection effect.
- Two UV-lamps
- Full user protection from direct UV radiation;
- Two operation modes — short time (under timer control) and continuous operation
- Digital control unit allows tracking of overall UV lamp operating time
- 1 min-24 hours/non stop timer
Recirculators fixation:
- Convenient fixation on walls (standard)
- Mounting on a movable tripod (optional)
Specifica
Specifica
| Filter Efficiency | 18 mW/cm2/sec, 14 m3/hour |
| Include | 2 Lamps |
| Montaggio | UVR-S |
| Product Type | UV-Air Flow Cleaner-Recirculator |