Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Codice prodotto. 12757839
Cambia vista
Click to view available options
Depth (Metric) Interior:
300 mm
325 mm
505 mm
690 mm
Altezza (sistema metrico) interno:
150 mm
200 mm
Larghezza (sistema metrico) interno:
150 mm
300 mm
Gamma di temperatura:
-15°C to +200°C
0°C to 200°C
Dimensione della confezione:
Pezzo
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Depth (Metric) Interior Altezza (sistema metrico) interno Larghezza (sistema metrico) interno Gamma di temperatura unitSize
12757839 505 mm 150 mm 300 mm 0°C to 200°C Pezzo
12747839 325 mm 150 mm 300 mm 0°C to 200°C Pezzo
12737839 300 mm 150 mm 150 mm 0°C to 200°C Pezzo
12767839 505 mm 200 mm 300 mm -15°C to +200°C Pezzo
12777839 690 mm 200 mm 300 mm -15°C to +200°C Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Codice prodotto. 12757839 Fornitore Grant Instruments™ N. del fornitore TX150ST18
Spedizione stimata: 05-10-2026
Aggiungi al carrello
Aggiungi al carrello
Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150°C. Grant Instruments™ Optima™ TX150 Series Digital Heated Circulating Bath, Stainless Steel Tank combines the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of five stainless steel (ST) tanks with different capacities.
  • Stability ±0.01ºC
  • Large, bright full colour display
  • Icon driven home screen via a dial and two function buttons
  • Programming/temperature profiling (TX150, 1 program with 30 segments)
  • Model available with/without clamp (T-Clamp)
  • Low-liquid detection (float switch)
  • 5 point user calibration
  • User adjustable over temperature dial
  • 5 point user calibration
TRUSTED_SUSTAINABILITY
Specifica
Gamma di temperatura 0°C to 200°C
Include Includes TX150 immersion heater/stirrer with ST18 Bath
Materiale Stainless Steel
Larghezza (sistema metrico) interno 300 mm
Altezza (sistema metrico) interno 150 mm
Depth (Metric) Interior 505 mm