Learn More
Grant Instruments™ Air Cleaner Recirculator, 1 Lamp
Descrizione
- Full user protection from direct UV radiation
- Simple to operate
- Convenient fixation on walls or mount on a movable tripod (optional)
- Compact, sturdy construction for everyday use
- Simple to operate with on/off switch
- UV lamp operation indicator – indicator light turns off when the lamp needs replacing
Applications:
Hospitals (outpatient departments, operating rooms, emergency rooms, delivery rooms etc.), kindergartens, research laboratories, veterinary clinics, schools, offices.
Specifications
Specifications
| Filter Efficiency | 18 mW/cm2/sec, 14 m3/hour |
| Include | 1 Lamp |
| Montaggio | UVR-S |
| Product Type | UV Cleaner-Recirculator |