Learn More
Abeba™ Air-Cushion Sole 2600 Shoes
Descrizione
- CE EN ISO 20347:2012 O2 SRA
- Grained leather
- White
- Slip resistance PU outsole sole
- Replaceable insole (article no. 3551)
- Sanitized treated
- Antistatic
- Lining microfiber, respiration absorbing
- Instep area with lacing
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 O2 SRA |
| Colore | White |
| Materiale rivestimento interno | Microfiber |
| Materiale | Leather |
| Size | 35 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 2600 |