Learn More
MedChemExpress Alisporivir intermediate-1
Specifica
Specifica
| CAS | 882506-05-0 |
| Content And Storage | Please store the product under the recommended conditions in the Certificate of Analysis. |
| Da utilizzare con (applicazione) | Neuroscience-Neuromodulation |
| Peso molecolare | 1461.91 g/mol |
| Product Type | Peptides |
| Quantità | 1 mg |
| Shipping Condition | Room temperature |