missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grace™ Alltech™ Vydac™ 218TP C18 Columns and Guard Cartridges
Click to view available options
Length (Metric):
250 mm
Diameter (Metric):
2.1 mm
3 mm
Particle Size:
5 μm
Unit Size:
Each
Description
- Acid and base deactivated symmetrical peaks
- Polymerically bonded and double-endcapped for long column lifetimes
- Analyze acids, bases, and neutrals in one run
Order Info
Guard Kits (Cat Nos. AT218GK104, -34, -54, -51 and -510) and Frits (AT218TP54T) are also available separately.
Specifications
Specifications
| Diameter (Metric) | 2.1 mm |
| Flow Rate | 1.2 mL/min |
| For Use With | Separation of Small Polypeptides <4000 to 5000 M.W., Enzymatic Digest Fragments, Natural and Synthetic Peptides, Multi-Ring Compounds |
| Length (Metric) | 250 mm |
| Mobile Phase Compatibility | A : 25% ACN/0.1% TFA aq. soln., B : 58% ACN/0.1% TFA aq. soln. |
| Particle Shape | Spherical |
| Particle Size | 5 μm |
| Pore Size | 300 Å |
| Surface Area | 60 to 110 m2/g |
| Temperature | 50°C |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction