Learn More
Allura Red AC (Technical Grade), TRC
Specifica
Specifica
| Materiale o nome chimico | Allura Red AC (E 129) |
| CAS | 25956-17-6 |
| Formula molecolare | C18 H14 N2 O8 S2 . 2 Na |
| Quantità | 1 g |
| Sinonimo | 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(2-methoxy-5-methyl-4-sulfophenyl)azo]-, disodium salt (9CI), 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(6-methoxy-4-sulfo-m-tolyl)azo]-, disodium salt (8CI), Allura Red 40, Allura Red AC, Allura red, C.I. 16035, C.I. Food Red 17, E 129, E 129 (dye), FD and C Red No. 40, FD&C Red No. 40, FDC Red 40, FDC Red 40 dye, Fancy Red, Food Red 17, Food Red No. 17, Food Red No. 40, Japan Food Red No. 40, Japan Red 40, Red 40, Red No. 40 |
| InChI Formula | InChI=1S/C18H16N2O8S2.2Na/c1-10-7-14(16(28-2)9-17(10)30(25,26)27)19-20-18-13-5-4-12(29(22,23)24)8-11(13)3-6-15(18)21;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;; |
| SMILES | [Na+].[Na+].COc1cc(c(C)cc1\N=N\c2c(O)ccc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC Name | disodium;6-hydroxy-5-[(E)-(2-methoxy-5-methyl-4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate |
| Molecular Weight (g/mol) | 496.42 |
| Peso formulazione | 495.9987 |