Learn More
Ansell AlphaTec™ 1500 PLUS Stitched Coveralls, Model 111, White
Product Code. 18000140 Shop All Ansell Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
6X-Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18000140 4X-Large Each
18080120 5X-Large Each
18070150 6X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18000140 Supplier Ansell Supplier No. WH15S0011108
Spedizione stimata: 24-06-2027
Provides comfortable protection from hazardous particulates in critical environments.
    • Specialized protection: The AlphaTec™ 1500 PLUS Stitched - Model 111 delivers light chemical resistance, guards against asbestos and filters 99.9% of particles over 3 microns in size, as well as brick and cement dust, light aerosols and mists
    • Heightened breathability: This asbestos protection suit's air and water vapor permeability also reduces wearers' exposure to heat stress
    • Enhanced fit and features: Wearer comfort is augmented by this protective body suit's optimal fit, while a three-piece hood offers additional protective coverage, finger loops offer a secure hold and red stitching ensures easy identification
    • Certified antistatic capabilities: The AlphaTec™ 1500 PLUS Stitched - Model 111 is tested against EN 1149-5 antistatic standards
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 4X-Large
Gender Unisex
Material Anti-static SMS
Color White
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Finger Loops, Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Chemicals, Particle
Certifications/Compliance (Europe) EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|CE Number|EN 1073-2:2002|EN 1149-5:2018|EN ISO 13688:2013
Latex Free Yes
For Use With (Application) Unpacking of bulk goods, Loading and unloading of proces equipment, Body Surface checking, marking and touching up, Assembly and inspection of components, Painting or cleaning tools or robots, Production line support and maintenance