Descrizione
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifica
Specifica
| Product Type | Chemical Resistant Lab Coat |
| Size | Small |
| Sesso | Unisex |
| Materiale | Microporous Polyethylene Laminate |
| Colore | White |
| Tipo di chiusura | Zipper |
| Stile polso | Elastic |
| Monouso | Limited-use |
| Sterilità | Non-sterile |
| Resistenza | Chemicals |
| Vedi altri risultati |