Learn More
Ansell™ AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped
Descrizione
- The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and taped - Model 214 chemical protective suit is made with a multi-layer barrier fabric, which offers wearers excellent protection against several chemicals, including inorganic and biological hazards
- These highly visible, bright yellow chemical protection coveralls improve worker safety in dimly lit environments
- Featuring welded and taped seams, this full chemical protection suit offers a strong barrier against harmful liquid and particles
Specifica
Specifica
| Tipo | Apron |
| Size | Medium |
| Materiale | Multi-layer Barrier Laminate Fabric |
| Colore | Yellow |
| Monouso | Single-use |
| Tipo di chiusura | Hook and Loop (Rear) |
| Sterilità | Non-sterile |
| Senza lattice | Yes |
| N. di tasche | None |
| Tipo di giuntura | Welded and Taped |
| Vedi altri risultati |