Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 756
Descrizione
- Specialized protection: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 chemical protective suit has proven to protect wearers from over 200 chemicals.
- Enhanced features: This full protective suit features a panoramic visor design that ensures an unobstructed field of vision
- Genuine versatility: The AlphaTec 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 full chemical protection suit also has an HEPA filter, which provides secondary protection from airline contamination and an SMC silencer keeps noise in the suit below 70 dB (even at maximum airflow)
- Considered aesthetics: In addition, the attached socks with elasticated boot overflap was designed to be worn inside chemical protective boots with the leg overflap worn outside, to reduce the potential for chemical ingress
- Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
- Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 AlphaTec™ 37-676 or G05 AlphaTec™ 29-500 gloves.
- Attached boot ends with anti-slip PVC sole version also available and emergency rip cord.
Specifica
Specifica
| Certificazione/Conformità | CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5) |
| Colore | Yellow |
| Da utilizzare con (applicazione) | Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment |
| Hood Type | Fully Encapsulated |
| Materiale | Multi-layer Barrier Laminate Fabric |
| Size | Small |
| Stile della caviglia | Elasticated Boot Overflap |
| Monouso | Reusable |
| Sesso | Unisex |
| Product Type | Encapsulated chemical protective suits for use with air-fed/supplied systems |
| Vedi altri risultati |