Learn More
Ansell AlphaTec™ 5000 APOLLO
Fully encapsulated chemical protective, liquid tight suit with Type 3/4 protection.
Marca:  Ansell OR50T-00186-07-G02
Dettagli aggiuntivi : Peso : 2.49200kg
Codice prodotto. 17110905
  • 342.64€ / Pezzo
Spedizione stimata: 14-04-2023
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Anti-static
Specifica
Specifica
Orange
Multi-Layer Non-Woven Barrier Laminate Fabric
Welded and Taped
132 gsm
Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases
3X-Large
O50T186B