Learn More
Ansell AlphaTec™ 5000 APOLLO
Catalog Number p-7228403
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Fully encapsulated chemical protective, liquid tight suit with Type 3/4 protection.
  • Outstanding permeation performance to numerous organic and inorganic chemicals, biological hazards and chemical warfare agents
  • Exceptional barrier protection, providing >480 minutes breakthrough time against 14 of 15 chemicals listed in EN ISO 6529
  • Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Anti-static
Colore Orange
Da utilizzare con (applicazione) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes, Unexpected Leakages Spills or Other Releases
Materiale Multi-Layer Non-Woven Barrier Laminate Fabric
Size Small
Tipo di giuntura Welded and Taped
Stile O50T186B
Peso (sistema metrico) 132 gsm