Learn More
Ansell™ AlphaTec™ Apron with Sleeves Ultrasonically Welded and Taped
Product Code. 31114783 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31114783 Large
31114781 Small
31114782 Medium
31114784 X-Large
31114785 2X-Large
31114786 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31114783 Supplier Ansell™ Supplier No. YE30T0021504
Spedizione stimata: 10-12-2026
Multi-layer barrier fabric, safeguards against numerous chemicals with comfort and durability assured
  • The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped - Model 215 is made with a multi-layer fabric, offering wearers protection against several inorganic chemicals and biological hazards
  • This highly visible, bright yellow chemical protective suit is designed to protect workers and enhance workplace safety
  • Made from laminate fabric, the AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded Taped - Model 215 ensures comfort and durability
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Seam Type Welded and Taped
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126)