Learn More
Ansell AlphaTec™ Sure Step Overshoes - Model 403
Product Code. 17150635
Change view
Click to view available options
Size:
46 to 48
Unit Size:
250 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17150635 46 to 48 250 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17150635 Supplier Ansell Supplier No. BLSSS0040305
Spedizione stimata: 23-06-2027
AlphaTec™ Sure Step Overshoes are disposable shoes that provides slip-resistance to the underside of the feet.
  • A disposable shoe that provides slip-resistance to the underside of the feet.
TRUSTED_SUSTAINABILITY
Specifications
Size 46 to 48
Material Polypropylene
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Certifications/Compliance (Europe) Not certified PPE
Resistance Chemical-resistant
Product Type Controlled Environments Shoe Covers