Learn More
Bryson Packaging™ Aluminum Foil Rolls
Product Code. 12359548
Change view
Click to view available options
Width (Metric):
300mm
450mm
600mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
12359548 300mm Each
12369548 450mm Each
12389548 600mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12359548 Supplier Bryson Packaging™ Supplier No. AKL300020P
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In stock
Add to basket
Edge
Add to basket
Ideal for use in autoclaves. Bryson Packaging™ Aluminum Foil Rolls are suitable for many laboratory applications including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes.
  • Available in a variety of widths
  • Can be cut to any length, cutter provided with box
  • Sealing, weighing and transferring samples
Specifications
Applications Ideal for use in autoclaves; 'Laboratory applications -including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes
Product Type Foil
Width (Metric) 300mm
Material Aluminum
Thickness 12 μm