Learn More
Korff™ Aluminum Foils
Product Code. 10136710
Change view
Click to view available options
Description:
10000 L x 500mm W
Unit Size:
10m
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10136710 10000 L x 500mm W 10m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10136710 Supplier Korff™ Supplier No. 10420
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
Spedizione stimata: 07-12-2026
Specifications
Description 10000 L x 500mm W
Product Type Foil
Width (Metric) 500mm
Material Aluminum
Thickness 50 μm