missing translation for 'onlineSavingsMsg'
Learn More

Ambroxol Hydrochloride, TRC

Codice prodotto. 17994085
Cambia vista
Click to view available options
Quantità:
100 mg
1 g
25 g
Dimensione della confezione:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17994085 1 g 1g
30134007 25 g 25g
17930755 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17994085 Fornitore Toronto Research Chemicals N. del fornitore TRCA5759001G
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 23828-92-4
Formula molecolare C13 H18 Br2 N2 O . Cl H
Molecular Weight (g/mol) 414.56
Sinonimo Cyclohexanol, 4-[(2-amino-3,5-dibromobenzyl)amino]-, hydrochloride, trans- (8CI), Cyclohexanol, 4-[[(2-amino-3,5-dibromophenyl)methyl]amino]-, monohydrochloride, trans- (9CI), Abramen, Ambril, Ambrodil, Ambron, Ambroxol hydrochloride, Bronchopront, Coughnol, Duramucal, Fluibron, Fluixol, Frenopect, Lindoxyl, Motosol, Mucasan, Muco-Burg, Mucoclear, Mucofar, Mucolear, Mucolin, Mucolite, Mucosal, Mucosolvan, Mucovent, N-(trans-4-Hydroxycyclohexyl)-2-amino-3,5-dibromobenzylamine hydrochloride, Pect, Solvolan, Stas-Hustenloser, Surbronc, Surfactal, trans-4-[(2-Amino-3,5-dibromobenzyl)amino]cyclohexanol hydrochloride
IUPAC Name 4-[(2-amino-3,5-dibromophenyl)methylamino]cyclohexan-1-ol;hydrochloride
SMILES Cl.Nc1c(Br)cc(Br)cc1CN[C@@H]2CC[C@@H](O)CC2

Specifica

Materiale o nome chimico Ambroxol hydrochloride
Note sul grado di purezza HPLC
Quantità 1 g
InChI Formula InChI=1S/C13H18Br2N2O.ClH/c14-9-5-8(13(16)12(15)6-9)7-17-10-1-3-11(18)4-2-10;/h5-6,10-11,17-18H,1-4,7,16H2;1H/t10-,11-;
Peso formulazione 411.9553
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.