Learn More
Aminoacetonitrile Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Aminoacetonitrile Hydrochloride |
| CAS | 6011-14-9 |
| Formula molecolare | C2H5ClN2 |
| Quantità | 10 g |
| Sinonimo | 2-Amino-acetonitrile Hydrochloride, Amino-acetonitrile Monohydrochloride, Glycinonitrile Monohydrochloride, 2-Aminoacetonitrile Hydrochloride, Aminoacetonitrile Hydrochloride, Aminoacetonitrile Monohydrochloride, Glycinonitrile Hydrochloride, alpha-Aminoacetonitrile Hydrochloride |
| InChI Formula | InChI=1S/C2H4N2.ClH/c3-1-2-4;/h1,3H2;1H |
| SMILES | Cl.NCC#N |
| IUPAC Name | 2-aminoacetonitrile;hydrochloride |
| Molecular Weight (g/mol) | 92.53 |
| Peso formulazione | 92.0141 |