missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Cerium(Iv) Nitrate, ≥98.0% (T), purum p.a., Honeywell™
Click to view available options
Quantità:
100 g
500 g
Imballaggio:
Poly Bottle
Descrizione
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifica
Materiale o nome chimico | Ammonium Cerium(Iv) Nitrate |
Quantità | 100 g |
Formula lineare | Ce(NH4)2(NO3)6 |
Numero UN | UN3085 |
Peso formulazione | 548.22g/mol |
Percent Purity | ≥98.0% (T) |
Grado | purum p.a. |
Imballaggio | Poly Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'