missing translation for 'onlineSavingsMsg'
Learn More

Ammonium metavanadate, Puriss. p.a., Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog Number p-7072001
Click to view available options
Quantità:
100 g
1 kg
50 kg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
vanadiumoylol amine
N.O[V](=O)=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
7803-55-6
H4NO3V
116.98
MFCD00011430
YBVKNHXQSRDWAA-UHFFFAOYSA-M
ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
516859
vanadiumoylol amine
N.O[V](=O)=O

Puriss. p.a., Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Ammonium Metavanadate
Nota nome Reag. Ph. Eur.
Quantità 100 g
Formula lineare NH4VO3
Numero UN 2859
Peso formulazione 116.98g/mol
Percent Purity ≥99.5%
Grado puriss. p.a.
Imballaggio Plastic Bottle
Densità 2.32g/cm3at 25°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'