missing translation for 'onlineSavingsMsg'
Learn More

Ammonium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Codice prodotto. 11380829
Cambia vista
Click to view available options
Quantità:
5 g
50 g
250 g
Dimensione della confezione:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11380829 50 g 50g
11370829 5 g 5g
11360829 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Normativa: Questo prodotto contiene un precursore di esplosivi ai sensi del regolamento (CE) n. 1148/2019. Ogni cliente deve compilare una dichiarazione prima della consegna.
Codice prodotto. 11380829 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 010638.18
 Articolo soggetto a restrizioni
Spedizione stimata: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

It is used in agriculture as a high-nitrogen fertilizer. It is one of the main components of ammonium nitrate/fuel oil (ANFO). ANFO is used in coal mining, quarrying, metal mining, and civil construction. It is also used as an oxidizer in solid rocket propellants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6484-52-2
Formula molecolare H4N2O3
Molecular Weight (g/mol) 80.04
Numero MDL MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Sinonimo ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O

Specifications

Punti di ebollizione 210°C (decomposition)
Materiale o nome chimico Ammonium nitrate
Punto di fusione 170°C
Colore Colorless
Forma fisica Crystalline
Quantità 50 g
Odore Odorless
Aspetto Colorless crystalline
Intervallo percentuale saggio 100.00%
Densità 1.72
Informazioni di solubilità Highly soluble in water; soluble in alcohol and liquid ammonia.
Peso formulazione 80.04
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.