missing translation for 'onlineSavingsMsg'
Learn More

Ampiroxicam, TRC

Codice prodotto. 30345755
Cambia vista
Click to view available options
Quantità:
10 g
Dimensione della confezione:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30345755 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30345755 Fornitore Toronto Research Chemicals N. del fornitore TRCA63435010G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Materiale o nome chimico Ampiroxicam
CAS 99464-64-9
Formula molecolare C20 H21 N3 O7 S
Quantità 10 g
InChI Formula InChI=1S/C20H21N3O7S/c1-4-28-20(25)30-13(2)29-18-14-9-5-6-10-15(14)31(26,27)23(3)17(18)19(24)22-16-11-7-8-12-21-16/h5-13H,4H2,1-3H3,(H,21,22,24)
SMILES CCOC(=O)OC(C)OC1=C(N(C)S(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3
IUPAC Name ethyl 1-[[2-methyl-1,1-dioxo-3-(pyridin-2-ylcarbamoyl)-1λ^{6},2-benzothiazin-4-yl]oxy]ethyl carbonate
Molecular Weight (g/mol) 447.46
Peso formulazione 447.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.