Learn More
Amsacrine Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | Amsacrine Hydrochloride |
| CAS | 54301-15-4 |
| Formula molecolare | C21H19N3O3S . HCl |
| Quantità | 50 mg |
| Sinonimo | N-[4-(9-acridinylamino)-3-methoxyphenyl]-Methanesulfonamide monohydrochloride (9CI), 4’-(Acridin-9-ylamino)-3'-methoxymethanesulfonanilide hydrochloride, AMSA hydrochloride, N-[4-(9-Acridinylamino)-3-methoxyphenyl]methanesulfonamide hydrochloride, NSC 141549, m-AMSA hydrochloride |
| InChI Formula | InChI=1S/C21H19N3O3S.ClH/c1-27-20-13-14(24-28(2,25)26)11-12-19(20)23-21-15-7-3-5-9-17(15)22-18-10-6-4-8-16(18)21;/h3-13,24H,1-2H3,(H,22,23);1H |
| SMILES | Cl.COc1cc(NS(=O)(=O)C)ccc1Nc2c3ccccc3nc4ccccc24 |
| IUPAC Name | N-[4-(acridin-9-ylamino)-3-methoxyphenyl]methanesulfonamide;hydrochloride |
| Molecular Weight (g/mol) | 429.9 |