Learn More
TFA Dostmann Analog Mini Weather Station
Codice prodotto. 15733055
Cambia vista
Click to view available options
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione della confezione
15733055 Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15733055 Fornitore TFA Dostmann N. del fornitore 20.3015.54
  • ONLINE EXCLUSIVE PRICE
    30.78€
    22.57€ / pezzi 1
    Risparmia 8.21€
    26% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 19-01-2027
The analog mini weather station, with a metallic finish plastic housing, is guaranteed to fit on any desk or shelf and it provides you with the latest data on temperature, air pressure,and humidity. The instrument is entirely manufactured in Germany. A compact instrument packed with information.
A compact instrument packed with information. The mini weather station, with a metallic finish plastic housing, is guaranteed to fit on any desk or shelf and it provides you with the latest data on temperature, air pressure (weather forecast) and humidity. The instrument is entirely manufactured in Germany.
  • Made in Germany
  • Table standing or wall mounting mini weather station
  • With barometer, thermometer and hygrometer
  • Barometer for local weather forecast
  • Monitoring of room temperature and humidity
Specifica
Accuracy (Barometer) ±8 mbar