Learn More
PepSlide™ Analyzer Perpetual License
Codice prodotto. 16347748
Cambia vista
Click to view available options
Software liscence
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
16347748 Software liscence
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16347748 Fornitore PepSlide™ N. del fornitore PSA.002.001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Intuitive and easy-to handle software for quantification of microarray data from PEPerCHIP™ Peptide Microarrays.
  • The software is adjusted to the PEPperCHIP™ Peptide Microarray platform and links microarray design files to content information for microarray data analysis
  • Software supports: quantification of microarray data, data analysis of pre-defined microarray content, microarray autoalignment, state-of-the-art noise and background correction algorithms and spot morphology recognition
Specifica
Tipo Software License
Da utilizzare con (applicazione) Quantification of flourescent intensities from PEPperCHIP Peptide Microarrays for antibody validation, epitope mapping, serum screening, biomarker discovery, protein-peptide interaction studies
Da utilizzare con (apparecchiatura) PEPperPRINT™ Peptide Microarrays