Learn More
Abeba™ Anatom 32171 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Marca: Abeba™ 32171/40
Dettagli aggiuntivi : Peso : 1.30000kg
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Microfiber black
- Steel toe cap
- Shock absorbant PU midsole
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3580)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Suitable for hygienically demanding areas
- Suitable for heel and foot lift
- Scratch-resistant coating in heel and toe area
Specifica
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Slip-resistant | |
32171 |
Black | |
40 | |
Polyurethane | |
Steel |