Learn More
ABEBA™ Anatom Green/Yellow Safety Shoes S3 ESD
Green/yellow Anatom shoes made of water/tear resistant Cordura, slip resistant PU2D outsole, steel toe cap, textile antipuncture
Marca: ABEBA™ 32251/47
Dettagli aggiuntivi : Peso : 1.33000kg
Descrizione
- Comfortable, robust, durable shoes
- Comply to EN ISO 20345: 2012, S3, SRC
- ESD dissipative according to EN 61340 class climatic 1
- Slip resistant PU double density outsole
- Green/yellow water and tear resistant Cordura
- Steel toe cap (200 J)
- Textile antipuncture midsole
- Scratch-resistant coated fabric in toe and heel area
- Breathable and antibacterial lining with silver ions
- Removable active comfort insole
Specifica
Green, Yellow | |
47 | |
EN ISO 20345: 2012 S3 SRC, ESD class 1 according EN 61340 | |
Low-cut | |
S1P SRC ESD |
Cordura | |
Steel | |
Men's | |
Water Resistant |