Learn More
Abcam Anisomycin, Protein Synthesis Inhibitor, Solid, 98% Purity, 10 mg
Product Code. 32173962 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32173962 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32173962 Supplier Abcam Supplier No. ab12049510mg
 Restricted Item
Spedizione stimata: 24-11-2026
Potent translational inhibitor of protein synthesis and activator of kinase cascades, including p38 and JNK MAP kinases.
  • Molecular Weight: 265.3 Da
  • Purity: >98%
  • Description: Translational inhibitor of protein synthesis. Highly potent activator of kinase cascades. Potently activates stress-activated protein kinases, p38, JNK MAP and other kinases.
  • Form: Solid
  • Storage: Store at +4°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule Protein, p38, JNK
Formulation 100% Anisomycin
Quantity 10 mg
For Use With (Application) Protein synthesis inhibition, Kinase cascade activation