Learn More
anti-ADRB2, Human, IP-WB Antibody Pair, Abnova™
Descrizione
Includes
Antibody pair for IP: rabbit polyclonal anti-ADRB2 (300μL), Antibody pair for WB: mouse purified polyclonal anti-ADRB2 (50μg)
Specifica
Specifica
| Antigene | ADRB2 |
| Applicazioni | Immunoprecipitation, Western Blot |
| Classificazione | Monoclonal/Polyclonal |
| Coniugato | Unconjugated |
| Descrizione | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | ADRB2 |
| Alias gene | ADRB2R/ADRBR/B2AR/BAR/BETA2AR |
| Simboli geni | ADRB2 |
| Specie ospite | Mouse, Rabbit |
| Quantità | 1 Set |
| Vedi altri risultati |
For Research Use Only