missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Adrenergic Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA533338
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon (95%) Guinea pig (90%) Monkey, Elephant, Rabbit (85%).
beta-3 Adrenergic Receptor Polyclonal antibody specifically detects beta-3 Adrenergic Receptor in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifica
beta-3 Adrenergic Receptor | |
Polyclonal | |
Unconjugated | |
ADRB3 | |
ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
155, 25645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P13945, P26255 | |
ADRB3 | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human ADRB3. | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |