missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-AGAP2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24498SS
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
AGAP2 Polyclonal specifically detects AGAP2 in Human, Mouse, Rat, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifica
| AGAP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| AGAP2 | |
| A portion of amino acids 450-500 of human AGAP2 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a),mouse, dog and monkey and 86% homologous in opossum. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| 0.5 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 ug/ml, Knockdown Validated | |
| AGAP-2, ArfGAP with GTPase domain, ankyrin repeat and PH domain 2, arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2, centaurin, gamma 1, centaurin-gamma-1, CENTG1, Cnt-g1, FLJ16430, GGAP2, GTP-binding and GTPase activating protein 2, GTP-binding and GTPase-activating protein 2, KIAA0167, Phosphatidylinositol-3-kinase enhancer, phosphoinositide 3-kinase enhancer, PIKEArf GAP with GTP-binding protein-like, ANK repeat and PH domains 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 116986 | |
| Human, Mouse, Rat, Equine, Primate | |
| IgG |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?
For Research Use Only